Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL10
Entrez Gene ID
102181157
Gene Name
kelch like family member 10
Gene Aliases
-
Chromosome Location
Chr.19: 41669933 - 41682907 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693830.2 XP_005693887.1 1 - 2 404 69

View Full Product Details