Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM17
Entrez Gene ID
102175411
Gene Name
transmembrane protein 17
Gene Aliases
-
Chromosome Location
Chr.11: 60687346 - 60695572 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686789.3 XP_005686846.2 2 - 3 324 123

View Full Product Details