Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDR9C7
Entrez Gene ID
102184503
Gene Name
short chain dehydrogenase/reductase family 9C member 7
Gene Aliases
-
Chromosome Location
Chr.5: 55922054 - 55931707 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013963934.2 XP_013819388.1 3 - 4 863 68

View Full Product Details