Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTRC
Entrez Gene ID
102174957
Gene Name
chymotrypsin C
Gene Aliases
-
Chromosome Location
Chr.16: 50536689 - 50543068 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690815.3 XP_005690872.1 6 - 7 674 92

View Full Product Details