Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC17
Entrez Gene ID
106501718
Gene Name
leucine rich repeat containing 17
Gene Aliases
-
Chromosome Location
Chr.4: 76027589 - 76068273 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013963247.2 XP_013818701.2 2 - 3 1227 115
GenBank mRNA JO498515.2 - 2 - 3 819 115

View Full Product Details