Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM72A
Entrez Gene ID
102174587
Gene Name
family with sequence similarity 72 member A
Gene Aliases
-
Chromosome Location
Chr.16: 3340268 - 3353200 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690410.3 XP_005690467.1 1 - 2 1030 114

View Full Product Details