Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPDR1
Entrez Gene ID
102175182
Gene Name
ependymin related 1
Gene Aliases
-
Chromosome Location
Chr.4: 70520395 - 70555461 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018047200.1 XP_017902689.1 1 - 2 608 80

View Full Product Details