Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HRC
Entrez Gene ID
102189986
Gene Name
histidine rich calcium binding protein
Gene Aliases
-
Chromosome Location
Chr.18: 56667530 - 56671702 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692705.3 XP_005692762.2 5 - 6 2362 68
GenBank mRNA JP350892.2 - 6 - 7 2056 68
JP350894.2 - 6 - 7 2071 68

View Full Product Details