Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CMC2
Entrez Gene ID
102179694
Gene Name
C-X9-C motif containing 2
Gene Aliases
-
Chromosome Location
Chr.18: 9018880 - 9033342 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018061757.1 XP_017917246.1 3 - 4 342 66
GenBank mRNA JO361973.2 - 1 - 2 371 66
JT760161.1 - 3 - 4 239 66

View Full Product Details