Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TXNDC17
Entrez Gene ID
102191830
Gene Name
thioredoxin domain containing 17
Gene Aliases
-
Chromosome Location
Chr.19: 25051460 - 25054030 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693429.3 XP_005693486.1 2 - 3 323 93

View Full Product Details