Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHM3
Entrez Gene ID
102187723
Gene Name
pleckstrin homology domain containing M3
Gene Aliases
-
Chromosome Location
Chr.2: 39616309 - 39830092 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005676438.3 XP_005676495.2 6 - 7 2405 102

View Full Product Details