Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPTC7
Entrez Gene ID
102180788
Gene Name
PTC7 protein phosphatase homolog
Gene Aliases
-
Chromosome Location
Chr.17: 16783834 - 16826233 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013970669.2 XP_013826123.2 3 - 4 964 90

View Full Product Details