Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZSCAN10
Entrez Gene ID
102184492
Gene Name
zinc finger and SCAN domain containing 10
Gene Aliases
-
Chromosome Location
Chr.25: 2419734 - 2423696 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018040195.1 XP_017895684.1 2 - 3 669 78

View Full Product Details