Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNJ13
Entrez Gene ID
102188013
Gene Name
potassium voltage-gated channel subfamily J member 13
Gene Aliases
-
Chromosome Location
Chr.3: 8291996 - 8299726 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005678736.3 XP_005678793.1 2 - 3 797 95
XM_018041112.1 XP_017896601.1 1 - 2 497 95
GenBank mRNA JO646492.2 - 1 - 2 468 95

View Full Product Details