Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MARCH3
Entrez Gene ID
102186006
Gene Name
membrane associated ring-CH-type finger 3
Gene Aliases
-
Chromosome Location
Chr.7: 83559110 - 83714160 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005682688.3 XP_005682745.1 4 - 5 803 81
XM_018050700.1 XP_017906189.1 3 - 4 695 81
GenBank mRNA JT619642.1 - 4 - 5 670 81

View Full Product Details