Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LALBA
Entrez Gene ID
100860779
Gene Name
lactalbumin alpha
Gene Aliases
ALA
Chromosome Location
Chr.5: 30812572 - 30814588 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285635.1 NP_001272564.1 1 - 2 146 91
GenBank mRNA EF564266.1 - 1 - 2 146 91
X05149.1 - 1 - 2 144 91

View Full Product Details