Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDF4
Entrez Gene ID
100860787
Gene Name
stromal cell derived factor 4
Gene Aliases
CAB45, SDF-4
Chromosome Location
Chr.16: 49770710 - 49783573 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285643.1 NP_001272572.1 2 - 3 442 76
XM_018059911.1 XP_017915400.1 3 - 4 603 76
XM_018059912.1 XP_017915401.1 3 - 4 611 76
XM_018059913.1 XP_017915402.1 3 - 4 579 76
GenBank mRNA EF589950.1 - 2 - 3 442 76
JO355186.2 - 3 - 4 627 76
JO500258.2 - 3 - 4 667 76

View Full Product Details