Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAM1
Entrez Gene ID
102169492
Gene Name
translocation associated membrane protein 1
Gene Aliases
TRAM
Chromosome Location
Chr.14: 47821760 - 47854029 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001314268.1 NP_001301197.1 5 - 6 546 87
XM_018058064.1 XP_017913553.1 5 - 6 1294 87
GenBank mRNA JO581586.1 - 6 - 7 1292 87
JQ308780.1 - 5 - 6 546 87
JQ308789.1 - 5 - 6 546 87

View Full Product Details