Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYM
Entrez Gene ID
100861166
Gene Name
prochymosin
Gene Aliases
Chy
Chromosome Location
Chr.3: 87617892 - 87628122 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285759.1 NP_001272688.1 8 - 9 1056 96
GenBank mRNA AY389343.1 - 8 - 9 1056 96
EF199763.1 - 8 - 9 1019 96
GQ369727.1 - 8 - 9 1007 96
JN377801.1 - 7 - 8 962 96

View Full Product Details