Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CASR
Entrez Gene ID
102185783
Gene Name
calcium sensing receptor
Gene Aliases
-
Chromosome Location
Chr.1: 66373772 - 66404333 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286092.1 NP_001273021.1 3 - 4 1571 67
XM_018059989.1 XP_017915478.1 3 - 4 1622 67
GenBank mRNA JO425752.1 - 3 - 4 1353 67
JO428925.2 - 3 - 4 1293 67
JT717514.1 - 4 - 5 1686 67
KF006348.1 - 3 - 4 1571 67

View Full Product Details