Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TNF
Entrez Gene ID
100861232
Gene Name
tumor necrosis factor
Gene Aliases
Cachectin, TNF-a, TNF-alpha, TNFA, TNFSF2
Chromosome Location
Chr.23: 22245929 - 22248692 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286442.1 NP_001273371.1 3 - 4 325 106
XM_005696606.3 XP_005696663.2 3 - 4 459 106
GenBank mRNA AF276985.1 - 3 - 4 157 106
AY304502.1 - 3 - 4 286 106
D86587.1 - 3 - 4 325 106
X14828.1 - 3 - 4 166 106

View Full Product Details