Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPLP0
Entrez Gene ID
102171363
Gene Name
ribosomal protein lateral stalk subunit P0
Gene Aliases
-
Chromosome Location
Chr.17: 8561763 - 8565823 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001314331.1 NP_001301260.1 2 - 3 225 90
XM_005709526.3 XP_005709583.3 3 - 4 316 90
GenBank mRNA JO581861.1 - 5 - 6 682 90
KM076930.1 - 2 - 3 225 90

View Full Product Details