Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBXL21P
Entrez Gene ID
102180122
Gene Name
F-box and leucine rich repeat protein 21, pseudogene
Gene Aliases
FBXL21
Chromosome Location
Chr.7: 63136718 - 63145648 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005682960.3 XP_005683017.2 3 - 4 847 90

View Full Product Details