Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PUS1
Entrez Gene ID
102175426
Gene Name
pseudouridine synthase 1
Gene Aliases
-
Chromosome Location
Chr.17: 26033777 - 26043972 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018061435.1 XP_017916924.1 3 - 4 876 103
XM_018061436.1 XP_017916925.1 3 - 4 461 103
XM_018061437.1 XP_017916926.1 3 - 4 499 103
GenBank mRNA JO423385.2 - 3 - 4 1052 103

View Full Product Details