Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BACH2
Entrez Gene ID
102173190
Gene Name
BTB domain and CNC homolog 2
Gene Aliases
-
Chromosome Location
Chr.9: 47246645 - 47636125 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018053143.1 XP_017908632.1 4 - 5 2067 93
XM_018053144.1 XP_017908633.1 5 - 6 2216 93
GenBank mRNA JT632420.1 - 7 - 8 2339 93

View Full Product Details