Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RWDD4
Entrez Gene ID
102187739
Gene Name
RWD domain containing 4
Gene Aliases
-
Chromosome Location
Chr.27: 31173310 - 31182955 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018042107.1 XP_017897596.1 2 - 3 361 82

View Full Product Details