Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APOBEC1
Entrez Gene ID
102181846
Gene Name
apolipoprotein B mRNA editing enzyme catalytic subunit 1
Gene Aliases
-
Chromosome Location
Chr.5: 100002861 - 100009443 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048507.1 XP_017903996.1 4 - 5 646 93

View Full Product Details