Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNMB3
Entrez Gene ID
102168667
Gene Name
potassium calcium-activated channel subfamily M regulatory beta subunit 3
Gene Aliases
-
Chromosome Location
Chr.1: 87446105 - 87551930 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013964515.2 XP_013819969.2 3 - 4 526 82
XM_018047560.1 XP_017903049.1 4 - 5 647 82
GenBank mRNA JO596546.2 - 3 - 4 485 82

View Full Product Details