Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BEAN1
Entrez Gene ID
102173221
Gene Name
brain expressed associated with NEDD4 1
Gene Aliases
-
Chromosome Location
Chr.18: 35470411 - 35509582 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018062083.1 XP_017917572.1 4 - 5 754 75

View Full Product Details