Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLAAT5
Entrez Gene ID
102176399
Gene Name
phospholipase A and acyltransferase 5
Gene Aliases
HRASLS5
Chromosome Location
Chr.29: 42095045 - 42113957 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043601.1 XP_017899090.1 5 - 6 853 76

View Full Product Details