Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLCL1
Entrez Gene ID
102174784
Gene Name
phospholipase C like 1 (inactive)
Gene Aliases
-
Chromosome Location
Chr.2: 49264010 - 49640327 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005676315.3 XP_005676372.2 1 - 2 733 94

View Full Product Details