Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RALA
Entrez Gene ID
102189928
Gene Name
RAS like proto-oncogene A
Gene Aliases
-
Chromosome Location
Chr.4: 38921824 - 38982755 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018046991.1 XP_017902480.1 3 - 4 612 108
GenBank mRNA JT657166.1 - 3 - 4 2077 108
KA344713.1 - 2 - 3 641 108

View Full Product Details