Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYL6
Entrez Gene ID
102170394
Gene Name
myosin light chain 6
Gene Aliases
-
Chromosome Location
Chr.5: 56536623 - 56539820 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005680364.3 XP_005680421.1 1 - 2 52 96

View Full Product Details