Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPP21
Entrez Gene ID
102189917
Gene Name
ribonuclease P/MRP subunit p21
Gene Aliases
-
Chromosome Location
Chr.23: 21535888 - 21537581 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018038839.1 XP_017894328.1 1 - 2 141 96

View Full Product Details