Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZCCHC13
Entrez Gene ID
102191025
Gene Name
zinc finger CCHC-type containing 13
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189516: 8523202 - 8524037 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005700621.2 XP_005700678.1 1 - 1 417 59

View Full Product Details