Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
YRDC
Entrez Gene ID
102189272
Gene Name
yrdC N6-threonylcarbamoyltransferase domain containing
Gene Aliases
-
Chromosome Location
Chr.3: 12693861 - 12697615 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018041898.1 XP_017897387.1 3 - 4 732 95
GenBank mRNA KA350362.1 - 5 - 6 1535 95

View Full Product Details