Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC9B
Entrez Gene ID
102186425
Gene Name
tetratricopeptide repeat domain 9B
Gene Aliases
-
Chromosome Location
Chr.18: 50508895 - 50514799 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018062411.1 XP_017917900.1 1 - 2 88 71

View Full Product Details