Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35G1
Entrez Gene ID
102184584
Gene Name
solute carrier family 35 member G1
Gene Aliases
-
Chromosome Location
Chr.26: 36204674 - 36214666 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018041632.1 XP_017897121.1 2 - 3 486 98
XM_018041634.1 XP_017897123.1 2 - 3 485 98
GenBank mRNA JO596459.2 - 2 - 3 191 98
KA338410.1 - 2 - 3 406 98

View Full Product Details