The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005682718.3 | XP_005682775.1 | 1 - 2 | 358 | 76 |
XM_018050670.1 | XP_017906159.1 | 1 - 2 | 358 | 76 |