Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LHFPL2
Entrez Gene ID
102189127
Gene Name
LHFPL tetraspan subfamily member 2
Gene Aliases
-
Chromosome Location
Chr.10: 91494172 - 91665917 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018053679.1 XP_017909168.1 4 - 5 923 57
XM_018053680.1 XP_017909169.1 5 - 6 1033 57
XM_018053681.1 XP_017909170.1 5 - 6 1026 57
XM_018053682.1 XP_017909171.1 4 - 5 1889 57
XM_018053683.1 XP_017909172.1 3 - 4 793 57
XM_018053684.1 XP_017909173.1 2 - 3 1081 57
XM_018053685.1 XP_017909174.1 2 - 3 1155 57
GenBank mRNA JT738801.1 - 1 - 2 466 57

View Full Product Details