Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DUSP2
Entrez Gene ID
102173395
Gene Name
dual specificity phosphatase 2
Gene Aliases
-
Chromosome Location
Chr.11: 2219090 - 2222726 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013967413.2 XP_013822867.2 1 - 2 1171 100
GenBank mRNA JO611331.2 - 3 - 4 1229 100

View Full Product Details