Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1H21orf91
Entrez Gene ID
102175802
Gene Name
chromosome 1 C21orf91 homolog
Gene Aliases
-
Chromosome Location
Chr.1: 17836540 - 17868813 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005674758.3 XP_005674815.2 4 - 5 935 75
XM_018051751.1 XP_017907240.1 4 - 5 830 75
GenBank mRNA JO329956.2 - 5 - 6 825 75
JO329958.2 - 5 - 6 825 75
JO355659.2 - 2 - 3 468 75
JO403437.2 - 1 - 2 320 75
JO455640.2 - 2 - 3 387 75
JO527946.3 - 1 - 2 96 75
JO532561.2 - 4 - 5 810 75
JO611771.2 - 1 - 2 544 75
JO611772.2 - 1 - 2 544 75
KA341029.1 - 4 - 5 805 75

View Full Product Details