Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAPS
Entrez Gene ID
102180118
Gene Name
calcyphosine
Gene Aliases
-
Chromosome Location
Chr.7: 92042219 - 92044844 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005682502.2 XP_005682559.1 3 - 4 1706 68
GenBank mRNA KA324844.1 - 1 - 2 561 68

Gene Symbol
RANBP3
Entrez Gene ID
102191605
Gene Name
RAN binding protein 3
Gene Aliases
-
Chromosome Location
Chr.7: 92044773 - 92095540 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JO646009.2 - 17 - 18 3541 68

View Full Product Details