Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JDP2
Entrez Gene ID
102172069
Gene Name
Jun dimerization protein 2
Gene Aliases
-
Chromosome Location
Chr.10: 16463235 - 16505507 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686097.3 XP_005686154.1 1 - 2 300 69

View Full Product Details