Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKX2-5
Entrez Gene ID
102180349
Gene Name
NK2 homeobox 5
Gene Aliases
-
Chromosome Location
Chr.20: 4963293 - 4966245 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013972667.2 XP_013828121.2 1 - 2 499 67

View Full Product Details