Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABT1
Entrez Gene ID
102179720
Gene Name
activator of basal transcription 1
Gene Aliases
-
Chromosome Location
Chr.23: 18454570 - 18458542 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013974100.2 XP_013829554.1 2 - 3 477 64
GenBank mRNA JO592717.2 - 1 - 2 773 64

View Full Product Details