Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GCC1
Entrez Gene ID
102171029
Gene Name
GRIP and coiled-coil domain containing 1
Gene Aliases
-
Chromosome Location
Chr.4: 28301710 - 28312280 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005679428.3 XP_005679485.2 1 - 2 1441 58
GenBank mRNA JT688348.1 - 3 - 4 1707 58

View Full Product Details