Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPACA9
Entrez Gene ID
102178283
Gene Name
sperm acrosome associated 9
Gene Aliases
-
Chromosome Location
Chr.11: 102356570 - 102370554 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018056046.1 XP_017911535.1 3 - 4 1162 77
GenBank mRNA JO574420.2 - 1 - 2 149 77

View Full Product Details