Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAMTOR1
Entrez Gene ID
102178377
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 1
Gene Aliases
-
Chromosome Location
Chr.15: 31697021 - 31703527 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018059411.1 XP_017914900.1 4 - 5 551 59
GenBank mRNA JO425085.1 - - 586 59
KA308179.1 - 1 - 2 644 59

View Full Product Details