Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZMAT1
Entrez Gene ID
102172520
Gene Name
zinc finger matrin-type 1
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189517: 41041721 - 41060738 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018044292.1 XP_017899781.1 2 - 3 833 93
XM_018044293.1 XP_017899782.1 2 - 3 617 93
XM_018044294.1 XP_017899783.1 4 - 5 488 93

View Full Product Details